Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium pyruvate, Hybri-Max™, powder, suitable for hybridoma
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium trichloroacetate, 97%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri™, ReagentPlus®, ≥99.5%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sign Into View Organizational & Contract Pricing SKU
Pack Size
Availability
Price
Quantity
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder