Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sign Into View Organizational & Contract Pricing Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sign Into View Organizational & Contract Pricing Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur™, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri™
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ACS reagent
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
5′-(N-Ethylcarboxamido)adenosine, powder
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sign Into View Organizational & Contract Pricing Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sign Into View Organizational & Contract Pricing Sigma-Aldrich
Adenosine, ≥99%
Sign Into View Organizational & Contract Pricing