MilliporeSigma
  • CD44 expression denotes a subpopulation of gastric cancer cells in which Hedgehog signaling promotes chemotherapy resistance.
CD44 expression denotes a subpopulation of gastric cancer cells in which Hedgehog signaling promotes chemotherapy resistance.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-06-21)
Changhwan Yoon, Do Joong Park, Benjamin Schmidt, Nicholas J Thomas, Hae-June Lee, Teresa S Kim, Yelena Y Janjigian, Deirdre J Cohen, Sam S Yoon
ABSTRACT
Gastric cancers may harbor a subset of cells with cancer stem cell (CSC) properties, including chemotherapy resistance, and CD44 is a gastric CSC marker. The Hedgehog (HH) pathway is a key developmental pathway that can be subverted by CSCs during tumorigenesis. Here, we examine the role of HH signaling in CD44(+) gastric cancer cells. Gastric cancer cell lines, tumor xenografts, and patient tumors were examined. Gastric cancer cell lines AGS, MKN-45, and NCI-N87 grown as spheroids or sorted for CD44(+) were found to have upregulation of HH pathway proteins. HH inhibition using Smoothened (Smo) shRNA or vismodegib (VIS) decreased spheroid formation and colony formation. CD44(+) cells, compared with unselected cells, were also resistant to 5-fluorouracil and cisplatin chemotherapy, and this resistance was reversed in vitro and in xenografts with Smo shRNA or VIS. CD44(+) cells also had significantly more migration, invasion, and anchorage-independent growth, and these properties could all be blocked with HH inhibition. Clinical tumor samples from a phase II trial of chemotherapy with or without VIS for advanced gastric cancer were analyzed for CD44 expression. In the chemotherapy alone group, high CD44 expression was associated with decreased survival, whereas in the chemotherapy plus VIS group, high CD44 expression was associated with improved survival. HH signaling maintains CSC phenotypes and malignant transformation phenotypes in CD44(+) gastric cancer cells, and HH inhibition can reverse chemotherapy resistance in CD44(+) cells. Gastric cancer is a heterogeneous disease, and the strategy of combining chemotherapy with HH inhibition may only be effective in tumors with high CD44 levels.
Sigma-Aldrich
MISSION® esiRNA, targeting human SMO
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DAPI, for nucleic acid staining
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
SKU
Pack Size
Availability
Price
Quantity
Cisplatin, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-SOX2 antibody produced in rabbit, IgG fraction of antiserum
SKU
Pack Size
Availability
Price
Quantity
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-sox2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-SOX2 antibody produced in rabbit