Merck
  • Accuracy of relative isotopic abundance and mass measurements in a single-stage orbitrap mass spectrometer.
Accuracy of relative isotopic abundance and mass measurements in a single-stage orbitrap mass spectrometer.
Rapid communications in mass spectrometry : RCM (2012-04-03)
A Kaufmann, S Walker
ABSTRACT
Orbitrap technology offers a combination of different technical specifications which have not yet been achieved by other high-resolution mass spectrometry instrumentation. This refers to the combination of sensitivity, dynamic range, mass accuracy, resolution and speed. The high stability of the mass axis and the general ease of use made the orbitrap instrumentation attractive for routine laboratories. However, there are circumstances where significantly deviating relative isotopic abundance (RIA) and shifting accurate masses can be observed. RIA becomes biased at low ion counts. Furthermore, two adjacent, only partially resolved near-isobaric ions are detected with a deviating RIA. The presence of a very intensive mass peak does not only induce Fourier transformation related artefacts (side-lobes) but can cause mass shifts of small adjacent near-isobaric mass peaks. These effects are not as drastic as known for Fourier transform ion cyclotron resonance instruments. Still, users trying to identify or quantify trace level compounds should be aware about such limitations in order to avoid possible pitfalls.
Sigma-Aldrich
tert-Butyl N-(2,3-dihydroxypropyl)carbamate, 97%