Merck
  • Novel bis(indolyl)maleimide pyridinophanes that are potent, selective inhibitors of glycogen synthase kinase-3.
Novel bis(indolyl)maleimide pyridinophanes that are potent, selective inhibitors of glycogen synthase kinase-3.
Bioorganic & medicinal chemistry letters (2007-03-14)
Han-Cheng Zhang, Llorente V R Boñaga, Hong Ye, Claudia K Derian, Bruce P Damiano, Bruce E Maryanoff
ABSTRACT
Novel bis(indolyl)maleimide pyridinophanes 3, 9a, 9b, 10a, 10b, and 11 were prepared by cobalt-mediated [2+2+2] cycloaddition of an appropriate alpha,omega-diyne with an N,N-dialkylcyanamide. These macrocyclic heterophanes were found to be potent, selective inhibitors of glycogen synthase kinase-3beta. An X-ray structure of a co-crystal of GSK-3beta and 3 (IC(50)=3nM) depicts the hydrogen bonding and hydrophobic interactions in the ATP-binding pocket of this serine/threonine protein kinase.
Sigma-Aldrich
6-Chloro-1-hexyne, 98%