- Human homologs of the Xenopus oocyte cortical granule lectin XL35.
Human homologs of the Xenopus oocyte cortical granule lectin XL35.
The cDNAs encoding two human homologs of the Xenopus oocyte lectin, XL35, were isolated from a small intestine cDNA library and termed HL-1 and HL-2. The deduced amino acid sequence of each homolog is about 60% identical and 80% similar to that of XL35, and none of these sequences contains the C-type lectin motif, although it is known that XL35 requires calcium for ligand binding. By Northern analysis, HL-1 transcripts are present at relatively high levels in heart, small intestine, colon, thymus, ovary, and testis. HL-2 transcripts, by contrast, are expressed only in small intestine. Immunocytochemistry using a polyclonal antibody produced against XL35 shows HL-1 protein to be localized exclusively in endothelial cells in colon, thymus, liver, and other tissues. Primary cultures of human aortic endothelial cells are positive for HL-1 expression by immunoblotting and by PCR analysis, but several other human cell types are not. HL-1 and -2 are both encoded at chromosome 1q23, the same locus that encodes the selectins. XL35, HL-1 and -2, and another mouse homolog are members of a new family of proteins whose members most likely perform diverse functions.