Merck
  • CpRu(PN) complex-catalyzed isomerization of allylic alcohols and its application to the asymmetric synthesis of muscone.
CpRu(PN) complex-catalyzed isomerization of allylic alcohols and its application to the asymmetric synthesis of muscone.
Journal of the American Chemical Society (2005-04-28)
Masato Ito, Sachiko Kitahara, Takao Ikariya
ABSTRACT
Highly efficient isomerization of allylic alcohols into saturated carbonyls is accomplished using the catalyst system of Cp*RuCl[Ph2P(CH2)2NH2-kappa2-P,N]-KOt-Bu (Cp* = eta5-C5(CH3)5) under mild conditions. Mechanistic consideration based on isotope-labeling experiments indicated the present reaction is applicable to the asymmetric isomerization of racemic sec-allylic alcohols with a prochiral olefin via dynamic kinetic resolution. A concise asymmetric synthesis of muscone has been achieved, where the asymmetric isomerization using an optically active ligand is a key reaction.
Sigma-Aldrich
α-Vinylbenzyl alcohol, 97%