Merck
  • Heterometallic CeIII-FeIII-salicylate networks: models for corrosion mitigation of steel surfaces by the 'green' inhibitor, Ce(salicylate)3.
Heterometallic CeIII-FeIII-salicylate networks: models for corrosion mitigation of steel surfaces by the 'green' inhibitor, Ce(salicylate)3.
Chemical communications (Cambridge, England) (2002-12-14)
Glen B Deacon, Craig M Forsyth, Thomas Behrsing, Kristina Konstas, Maria Forsyth
ABSTRACT
The syntheses and structures of the novel Ce-Fe bimetallic complexes [[Fe(sal)2(bpy)]2Ce(NO3)(H2O)3].EtOH and [[Fe(sal)2(bpy)]4Ce2(H2O)11][salH]2.EtOH.3H2O (salH2 = salicylic acid) suggest Fe(3+)-sal2- units and Ce-OC(R)O-Fe bridging contribute to the formation of corrosion inhibitive layers on steel surfaces exposed to [Ce(salH)3(H2O)].