- Expression of glial fibrillary acidic protein (GFAP) in peripheral nerve sheath tumors. A comparative study of immunoreactivity of GFAP, vimentin, S-100 protein, and neurofilament in 38 schwannomas and 18 neurofibromas.
Expression of glial fibrillary acidic protein (GFAP) in peripheral nerve sheath tumors. A comparative study of immunoreactivity of GFAP, vimentin, S-100 protein, and neurofilament in 38 schwannomas and 18 neurofibromas.
The American journal of surgical pathology (1988-02-01)
E Kawahara, Y Oda, A Ooi, S Katsuda, I Nakanishi, S Umeda
PMID3124642
ABSTRACT
Immunoreactivity of glial fibrillary acidic protein (GFAP) in 38 schwannomas and 18 neurofibromas was evaluated and compared with the reactivity of vimentin, S-100 protein, and neurofilament protein. All cases were positive for vimentin and S-100 protein. GFAP was positively stained in the neoplastic cells of 15 of 38 schwannomas (38%) and in two of 18 neurofibromas (11%). The extensively stained GFAP-positive tumors tended to be deeply situated in the body. The GFAP-positive cells were usually spindle-shaped and appeared preferentially in the perivascular region of hyalinized, thick blood vessels.