- Changes in cell surface antigen expression during hemopoietic differentiation.
Changes in cell surface antigen expression during hemopoietic differentiation.
Human bone marrow cells were separated on a fluorescence activated cell sorter (FACS) according to their binding of a series of monoclonal antibodies; the positive and negative fractions were cloned for erythroid burst and colony-forming units (BFU-E and CFU-E) and myeloid colony-forming units (CFU-GM), and cytocentrifuge slides were prepared for microscopy of maturing precursors. The pattern of antigen expression on hemopoietic progenitor and precursor populations has been established using antibodies defining blood group (A, I/i), HLA-associated (*A, B, C, DR, DC1), lineage specific, and transferrin receptor antigens. Like monomorphic HLA-DR, the antigen defined by monoclonal antibody OKT10 is expressed on the earliest progenitors and lost during differentiation, suggesting a role in interactions regulating the differentiation of these cells. The HLA-linked DC1 determinant, in contrast to HLA-DR, is not expressed at a detectable level on progenitor cells. Although a lineage-specific early antigen has not been identified, the transferrin receptor is expressed on the majority of erythroid progenitors, but only weakly on myeloid progenitors, and may provide an approach to isolating erythroid progenitors. These and earlier studies with monoclonal antibodies against HLA-DR and glycophorin now provide a detailed "map" of antigen expression during hemopoietic differentiation.