Merck
  • DNA damage and repair of human skin keratinocytes concurrently exposed to pyrene derivatives and UVA light.
DNA damage and repair of human skin keratinocytes concurrently exposed to pyrene derivatives and UVA light.
Toxicology research (2014-06-06)
Tracie Perkins Fullove, Hongtao Yu
ABSTRACT
Polycyclic aromatic hydrocarbons (PAHs), a class of mutagenic environmental contaminants, insert toxicity through both metabolic activation and light irradiation. Pyrene, one of the most widely studied PAHs, along with its mono-substituted derivatives, 1-amino, 1-bromo, 1-hydroxy, and 1-nitropyrene, were chosen to study the effect of substituents on their phototoxicity, DNA damage and repair. Both alkaline Comet assay, which detects direct DNA damages, and
Sigma-Aldrich
1-Bromopyrene, 96%