Merck
  • Aspirin therapy in primary cardiovascular disease prevention: a position paper of the European Society of Cardiology working group on thrombosis.
Aspirin therapy in primary cardiovascular disease prevention: a position paper of the European Society of Cardiology working group on thrombosis.
Journal of the American College of Cardiology (2014-07-19)
Sigrun Halvorsen, Felicita Andreotti, Jurriën M ten Berg, Marco Cattaneo, Sergio Coccheri, Roberto Marchioli, João Morais, Freek W A Verheugt, Raffaele De Caterina
ABSTRACT
Although the use of oral anticoagulants (vitamin K antagonists) has been abandoned in primary cardiovascular prevention due to lack of a favorable benefit-to-risk ratio, the indications for aspirin use in this setting continue to be a source of major debate, with major international guidelines providing conflicting recommendations. Here, we review the evidence in favor and against aspirin therapy in primary prevention based on the evidence accumulated so far, including recent data linking aspirin with cancer protection. While awaiting the results of several ongoing studies, we argue for a pragmatic approach to using low-dose aspirin in primary cardiovascular prevention and suggest its use in patients at high cardiovascular risk, defined as ≥2 major cardiovascular events (death, myocardial infarction, or stroke) projected per 100 person-years, who are not at increased risk of bleeding.
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard