Merck
  • Aggregation-induced emission, self-assembly, and electroluminescence of 4,4'-bis(1,2,2-triphenylvinyl)biphenyl.
Aggregation-induced emission, self-assembly, and electroluminescence of 4,4'-bis(1,2,2-triphenylvinyl)biphenyl.
Chemical communications (Cambridge, England) (2010-01-21)
Zujin Zhao, Shuming Chen, Xiaoyuan Shen, Faisal Mahtab, Yong Yu, Ping Lu, Jacky W Y Lam, Hoi Sing Kwok, Ben Zhong Tang
ABSTRACT
Two is better than one: a luminogen comprised of two units of tetraphenylethene (BTPE) emits more efficiently than that with one tetraphenylethene unit in the solid state; self-assembly of the BTPE molecules affords crystalline microfibers that fluoresce in 100% efficiency, giving the largest effect of aggregation-induced emission (alpha(AIE)-->infinity); BTPE-based electroluminescence devices emit in current efficiency up to approximately 7.3 cd/A.