Merck
  • The rationale of targeting neutrophils with dapsone during glioblastoma treatment.
The rationale of targeting neutrophils with dapsone during glioblastoma treatment.
Anti-cancer agents in medicinal chemistry (2011-06-29)
Richard E Kast, Angelika Scheuerle, Christian R Wirtz, Georg Karpel-Massler, Marc-Eric Halatsch
ABSTRACT
Data from past research is presented showing that neutrophils are active participants in new vessel formation in normal physiology, in proliferating human endometrium, in non-cancer pathologies as in the pannus of rheumatoid arthritis, and in various cancers, among them glioblastoma. These data show that interleukin-8 (IL-8) is a major chemokine attracting neutrophil infiltrates in these states. Since the old anti-Hansen's disease drug dapsone inhibits neutrophil migration along an IL-8 gradient towards increasing concentrations, and is used therapeutically for this attribute to good effect in dermatitis herpetiformis, bullous pemphigoid and rheumatoid arthritis, we suggest dapsone may deprive glioblastoma of neutrophil-mediated growth promoting effects. We review past research showing that vascular endothelial growth factor, VEGF, is carried predominantly intracellularly within neutrophils--only 2% of circulating VEGF is found free in serum. Based on the available evidence summarized by the authors, dapsone has a strong theoretical potential to become a useful anti-VEGF, anti-angiogenic agent in glioblastoma treatment.
Product Description
Supelco
Dapsone, VETRANAL®, analytical standard
Dapsone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-Aminophenyl sulfone, 97%