Merck
  • [An overview of the pharmacokinetics and pharmacodynamics of rocuronium bromide].
[An overview of the pharmacokinetics and pharmacodynamics of rocuronium bromide].
Masui. The Japanese journal of anesthesiology (2006-07-22)
Hiroshi Iwasaki
ABSTRACT
For many years, much effort has been done to develop non-depolarizing neuromuscular blocking agents of rapid onset and short duration. It has been shown that rocuronium bromide is a nondepolarizing, steroidal muscle relaxant with fast onset and short-to-intermediate duration of action. In addition, rocuronium-induced neuromuscular blockade can be readily and consistently reversed following the administration of reversal agents. The pharmacology and pharmacodynamics of rocuronium bromide are summarized in this review.
Sigma-Aldrich
Rocuronium bromide, ≥97% (perchloric acid titration)
Rocuronium bromide, European Pharmacopoeia (EP) Reference Standard
Rocuronium for peak identification, European Pharmacopoeia (EP) Reference Standard