Merck
  • Evaluation of testosterone/epitestosterone ratio influential factors as determined in doping analysis.
Evaluation of testosterone/epitestosterone ratio influential factors as determined in doping analysis.
Journal of analytical toxicology (2000-03-25)
D H van de Kerkhof, D de Boer, J H Thijssen, R A Maes
ABSTRACT
The ratio of the concentration of testosterone glucuronide to the concentration of epitestosterone glucuronide (T/E ratio) as determined in urine is the most frequently used method to prove testosterone abuse by athletes. A T/E ratio higher than 6 has been considered as proof of abuse in the past; however, cases of naturally occurring higher T/E ratios have been described. Since the introduction of the T/E ratio in doping analysis, the parameters that may or may not influence the T/E ratio, possibly leading to false-positive results, have been debated. To achieve more insight on the influencing circumstances, an overview is given to obtain an objective view on the merits of the urinary T/E ratio. Relevant analytical aspects of the T/E ratio, potential parameters of endogenous and exogenous origins, as well as some alternative methods to determine testosterone abuse, such as the urinary testosterone/luteinizing hormone ratio, gas chromatography-combustion-isotope-ratio mass spectrometry, hair analysis, and high-performance liquid chromatography-mass spectrometry, are discussed.
Product Description
Supelco
Epitestosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.