Merck
Assay development for histone methyltransferases.
Assay and drug development technologies (2013-04-06)
Kurumi Y Horiuchi, Mia M Eason, Joseph J Ferry, Jamie L Planck, Colin P Walsh, Robert F Smith, Konrad T Howitz, Haiching Ma
ABSTRACT
Epigenetic modifications play a crucial role in human diseases. Unlike genetic mutations, however, they do not change the underlying DNA sequences. Epigenetic phenomena have gained increased attention in the field of cancer research, with many studies indicating that they are significantly involved in tumor establishment and progression. Histone methyltransferases (HMTs) are a large group of enzymes that specifically methylate protein lysine and arginine residues, especially in histones, using S-adenosyl-L-methionine (SAM) as the methyl donor. However, in general, HMTs have no widely accepted high-throughput screening (HTS) assay format, and reference inhibitors are not available for many of the enzymes. In this study, we describe the application of a miniaturized, radioisotope-based reaction system: the HotSpot(SM) platform for methyltransferases. Since this platform employs tritiated SAM as a cofactor, it can be applied to the assay of any HMT. The key advantage of this format is that any substrate can be used, including peptides, proteins, or even nucleosomes, without the need for labeling or any other modifications. Using this platform, we have determined substrate specificities, characterized enzyme kinetics, performed compound profiling for both lysine and arginine methyltransferases, and carried out HTS for a small-library LOPAC against DOT1L. After hit confirmation and profiling, we found that suramin inhibited DOT1L, NSD2, and PRMT4 with IC₅₀ values at a low μM range.
Sigma-Aldrich
LOPAC®1280 - Small Scale
Sigma-Aldrich
LOPAC®1280 - Small Scale, International Version
Sigma-Aldrich
LOPAC®1280
Sigma-Aldrich
LOPAC®1280 (International Version)