Merck
  • First example of high loaded polymer-stabilized nanoclusters immobilized on hydrotalcite: effects in alkyne hydrogenation.
First example of high loaded polymer-stabilized nanoclusters immobilized on hydrotalcite: effects in alkyne hydrogenation.
Chemical communications (Cambridge, England) (2004-04-08)
Jules C A A Roelofs, Peter H Berben
ABSTRACT
Poly(N-vinyl-2-pyrrolidone)-stabilized Pd-nanoclusters, for the first time exclusively supported on the hydrotalcite lateral surface, showed a remarkable catalytic performance in the selective hydrogenation of 3-hexyn-1-ol, which can be ascribed to both the influence of the protecting polymer PVP as well as the nature of the support.
Sigma-Aldrich
3-Hexyn-1-ol, 98%