Merck
An organic solvent and surfactant stable α-amylase from soybean seeds.
Acta biochimica Polonica (2013-09-26)
Nivedita Jaiswal, Om Prakash
ABSTRACT
An organic solvent and surfactant stable α-amylase was obtained from soybean seeds. The direct and indirect effect of various organic solvents (non-polar, polar protic, and polar aprotic) and surfactants on the activity and stability of free enzyme was determined. The enzyme showed a very high catalytic efficiency and stabilization against most of the organic solvents and surfactants tested, except for few. Those organic solvents and surfactants (like chloroform, dimethyl formamide, n-butanol, and Tween 20), which caused an inhibition in enzyme activity, were used to study their effects on immobilized enzyme. The inhibitory effect was found to be decreased in immobilized enzyme as compared to free enzyme indicating that immobilization imparted stability to the enzyme. Moreover, the possibility of reuse of the enzyme in the presence of the organic solvents and surfactants was increased upon immobilization. The stability of soybean α-amylase towards organic solvents and surfactants shows that it is a potential candidate for use in organic-solvent biocatalysis as well as in detergent industries.
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)