Merck
  • Hyaluronidases and their inhibitors in the serum of colorectal carcinoma patients.
Hyaluronidases and their inhibitors in the serum of colorectal carcinoma patients.
Journal of pharmaceutical and biomedical analysis (2013-06-20)
C Kolliopoulos, D Bounias, H Bouga, D Kyriakopoulou, M Stavropoulos, D H Vynios
ABSTRACT
Colorectal cancer is the third most commonly diagnosed type of cancer. Hyaluronan is involved in this malignancy. Moreover, hyaluronidases - its degrading enzymes - display controversial roles regarding their involvement in tumor development. HYAL-1 is the major tumor derived hyaluronidase. The aim of the study was the determination and evaluation of hyaluronidase levels in serum of colorectal cancer patients, before and after surgery, with a view to assessing its potential role as a tumor marker for recurrence. By zymography and Western blotting, it was confirmed that HYAL-1 was the only hyaluronidase present in samples. Quantification of its activity indicated a statistically significant decrease in samples seven days postoperatively, compared with the respective ones before surgery. HYAL-1 levels before surgery were significantly reduced in comparison with healthy samples and samples one year postoperatively. Hyaluronidase inhibitor activity was demonstrated under mild alkaline conditions via reverse zymography. A statistically significant increase was observed in samples seven days postoperatively, when compared with samples before surgery. HYAL-1 levels in sera of colorectal cancer patients were lower than those of healthy population, possibly because of the local accumulation of the enzyme in tumor microenvironment. A gradual elevation up to one year postoperatively to reach healthy levels might indicate a role of HYAL-1 levels in cancer.
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid