Merck
  • Evaluation of passive samplers with neutral or ion-exchange polymer coatings to determine freely dissolved concentrations of the basic surfactant lauryl diethanolamine: Measurements of acid dissociation constant and organic carbon-water sorption coefficient.
Evaluation of passive samplers with neutral or ion-exchange polymer coatings to determine freely dissolved concentrations of the basic surfactant lauryl diethanolamine: Measurements of acid dissociation constant and organic carbon-water sorption coefficient.
Journal of chromatography. A (2013-10-08)
Fang Wang, Yi Chen, Joop L M Hermens, Steven T J Droge
ABSTRACT
A passive sampler tool (solid-phase microextraction, SPME) was optimized to measure freely dissolved concentrations (Cw,free) of lauryl diethanolamine (C12-DEA). C12-DEA can be protonated and act as a cationic surfactant. From the pH-dependent sorption to neutral SPME coatings (polyacrylate and PDMS), a pKa of 8.7 was calculated, which differs more than two units from the value of 6.4 reported elsewhere. Polyacrylate coated SPME could not adequately sample largely protonated C12-DEA in humic acid solutions of pH 6. A new hydrophobic SPME coating with cation-exchange properties (C18/SCX) sorbed C12-DEA 100 fold stronger than polyacrylate, because it specifically sorbs protonated C12-DEA species. The C18/SCX-SPME fiber showed linear calibration isotherms in a concentration range of <1 nM-1 μM (well below the CMC). Using the C18/SCX-SPME fibers, linear sorption isotherms to Aldrich humic acid at pH 6 (ionic strength 0.015 M) were measured over a broad concentration range with a sorption coefficient of 10(5.3).
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder