- [Aneurysm of the humeral artery].
[Aneurysm of the humeral artery].
Angiologia (1991-05-01)
R Romero Campos, M L Abellán Cubel, M Lerma Goncé, L Camacho
PMID1952257
ABSTRACT
Authors report the case of a young man affected by an aneurysm at the humeral artery. The relevance of this case was due to its notable rare incidence at such level as well as its difficult etiologic diagnosis. Patient was treated by an aneurysmectomy and a basilic vein ipsilateral was interposed. Immediate and after 18 months results were satisfactory. Arteriographic and anatomopathologic studies are presented.
Sigma-Aldrich
Fructosyl-Amino Acid Oxidase from Corynebacterium sp., recombinant, expressed in E. coli, lyophilized powder, ≥0.45 units/mg protein