Merck
  • Efficient strategy for the synthesis of stereopentad subunits of scytophycin, rifamycin S, and discodermolide.
Efficient strategy for the synthesis of stereopentad subunits of scytophycin, rifamycin S, and discodermolide.
Organic letters (2001-12-12)
S BouzBouz, J Cossy
ABSTRACT
[reaction: see text] An efficient, simple method has been developed for the stereocontrolled synthesis of polypropionate stereopentads in high enantio- and diastereomeric purities.
Product Description
Rifamycin S, European Pharmacopoeia (EP) Reference Standard