Merck
A transition in brain state during propofol-induced unconsciousness.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-01-17)
Eran A Mukamel, Elvira Pirondini, Behtash Babadi, Kin Foon Kevin Wong, Eric T Pierce, P Grace Harrell, John L Walsh, Andres F Salazar-Gomez, Sydney S Cash, Emad N Eskandar, Veronica S Weiner, Emery N Brown, Patrick L Purdon
ABSTRACT
Rhythmic oscillations shape cortical dynamics during active behavior, sleep, and general anesthesia. Cross-frequency phase-amplitude coupling is a prominent feature of cortical oscillations, but its role in organizing conscious and unconscious brain states is poorly understood. Using high-density EEG and intracranial electrocorticography during gradual induction of propofol general anesthesia in humans, we discovered a rapid drug-induced transition between distinct states with opposite phase-amplitude coupling and different cortical source distributions. One state occurs during unconsciousness and may be similar to sleep slow oscillations. A second state occurs at the loss or recovery of consciousness and resembles an enhanced slow cortical potential. These results provide objective electrophysiological landmarks of distinct unconscious brain states, and could be used to help improve EEG-based monitoring for general anesthesia.
Product Description
Supelco
Propofol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard
Propofol, European Pharmacopoeia (EP) Reference Standard