Merck
  • Characterization and comparative analysis of psychrophilic and mesophilic alpha-amylases from Euplotes species: a contribution to the understanding of enzyme thermal adaptation.
Characterization and comparative analysis of psychrophilic and mesophilic alpha-amylases from Euplotes species: a contribution to the understanding of enzyme thermal adaptation.
Biochemical and biophysical research communications (2013-08-07)
Guang Yang, Guang Yang, Lino Aprile, Vincenzo Turturo, Sandra Pucciarelli, Stefania Pucciarelli, Cristina Miceli
ABSTRACT
The eukaryotic α-amylase isolated from the psychrophilic ciliated protozoon Euplotes focardii (EfAmy) was expressed in Escherichia coli and biochemically characterized. Its enzymatic activity was compared to that of the homologous protein from the mesophilic congeneric species Euplotes crassus (EcAmy). The comparison of the amino acid composition and the surface residue composition of the two enzymes indicated a preference for tiny residues and the avoidance of charged, aromatic and hydrophobic residues in EfAmy. Our comparative homology modeling study reveals a lack of surface salt bridges, a decreased number of the surface charged residues, decreased hydrogen bonds and bound ions, and a reduction of aromatic-sulfur interactions, cationic-π interactions and disulfide interactions in EfAmy. In contrast, sequence alignment and homology modeling showed five unconserved prolines located on the surface loops of EcAmy. By analyzing amylolytic activity towards soluble starch as the substrate, we determined the temperature and pH dependence, thermostability and kinetic parameters of these two enzymes. We demonstrated that EfAmy shows the characteristics of a psychrophilic α-amylase, such as the highest hydrolytic activity at low temperatures and high thermolability. In contrast, the EcAmy showed mesophilic characteristics with the highest activity at moderate temperatures and a more than 2-fold increased half-life at 50°C compared to EfAmy. The kcat and KM values of EfAmy were higher than those of the mesophilic EcAmy at all tested temperatures. Furthermore, both EfAmy and EcAmy showed maximum activities at pH 9 and maintained high activities in the presence of surfactants. These results suggest the potential applications of EfAmy and EcAmy as ingredients in detergents for industrial applications.
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)