Merck
  • Enzyme kinetics and substrate selectivities of rat glutathione S-transferase isoenzymes towards a series of new 2-substituted 1-chloro-4-nitrobenzenes.
Enzyme kinetics and substrate selectivities of rat glutathione S-transferase isoenzymes towards a series of new 2-substituted 1-chloro-4-nitrobenzenes.
Xenobiotica; the fate of foreign compounds in biological systems (1996-02-01)
E M van der Aar, D Buikema, J N Commandeur, J M te Koppele, B van Ommen, P J van Bladeren, N P Vermeulen
ABSTRACT
1. Four different rat glutathione S-transferase (GST) isoenzymes, belonging to three different classes, were examined for their GSH conjugating capacity towards 11 2-substituted 1-chloro-4-nitrobenzene derivatives. Significant differences were found in their enzyme kinetic parameters Km, kcat and kcat/Km. 2. Substrates with bulky substituents on the ortho-position appeared to have high affinities (low Km's) for the active site of the GST-isoenzymes, suggesting that there is sufficient space in this area of the active site. A remarkably high Km (low affinity) was found for 2-chloro-5-nitropyridine towards all GST-isoenzymes examined. 3. GST 3-3 catalysed the reaction between GSH and the substrates most efficiently (high kcat) compared with the other GST-isoenzymes. Moreover, GST 3-3 showed clear substrate selectivities towards the substrates with a trifluoromethyl-, chlorine- and bromine-substituent. 1-Chloro-2,4-dinitrobenzene and 2-chloro-5-nitrobenzonitrile were most efficiently conjugated by all four GST-isoenzymes examined. 4. When the rate of the conjugation reactions was followed, a linear increase of formation of GS-conjugate could be seen for 2-chloro-5-nitrobenzonitrile during a much longer period of time than for 1-chloro-2,4-dinitrobenzene with all GST-isoenzymes examined. Therefore, it is suggested that 2-chloro-5-nitrobenzonitrile might be recommended as an alternative model substrate in GST-research.
Sigma-Aldrich
2-Chloro-5-nitrobenzonitrile, 99%