Merck
  • The role of cytochrome b5 in the biosynthesis of androgens by human P450c17.
The role of cytochrome b5 in the biosynthesis of androgens by human P450c17.
Archives of biochemistry and biophysics (1995-03-10)
M Katagiri, N Kagawa, M R Waterman
ABSTRACT
Human cytochrome b5 has a profound effect on the 17,20-lyase activities catalyzed by purified, human cytochrome P450c17. It enhances the conversion of 17 alpha-hydroxypregnenolone to dehydroepiandrosterone by 13-fold and the conversion of 17 alpha-hydroxyprogesterone to androstenedione by at least 10-fold. This latter activity is virtually undetectable in the absence of cytochrome b5. Other activities catalyzed by P450c17 include 17 alpha-hydroxylation of progesterone and pregnenolone and are much less influenced by cytochrome b5. The conversion of pregnenolone to 17 alpha-hydroxypregnenolone is increased by 2-fold, while that of progesterone to 17 alpha-hydroxyprogesterone is unchanged. These studies using purified systems suggest that cytochrome b5 plays a role in regulating the activities of P450c17 to optimize the balance between sex hormone synthesis and glucocorticoid synthesis. In particular, they indicate that in human testes which contains a high b5/P450 ratio, 17 alpha-hydroxyprogesterone can serve as an intermediate in testosterone production, rather than being a dead-end product, or stated another way, because of the relatively high concentration of cytochrome b5 in the human testis, both the delta 4 and the delta 5 steroidogenic pathways can lead to testosterone production.
Sigma-Aldrich
17α-Hydroxyprogesterone, ≥95%
SKU
Pack Size
Availability
Price
Quantity
Supelco
17α-Hydroxyprogesterone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®