Merck
  • Fatal opiates overdose. Toxicological identification of various metabolites in a blood sample by GC-MS after silylation.
Fatal opiates overdose. Toxicological identification of various metabolites in a blood sample by GC-MS after silylation.
Forensic science international (1998-08-26)
M Balíková, V Maresová
ABSTRACT
A fatal opiates overdose, where ethylmorphine, hydrocodone, dihydrocodeine and codeine were consumed concomitantly, is reported. This case report may contribute to data on fatal blood concentrations of drugs with rare incidence. The relative retention times in capillary gas chromatography and full mass spectra of various opiates in their silylated forms, detected together in one sample, may serve as a helpful analytical reference for clinical and forensic toxicologists.
Product Description
Supelco
Ethylmorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®