Merck
  • Somatostatin receptor-effector system in rat pancreatic acinar membranes after subtotal enterectomy.
Somatostatin receptor-effector system in rat pancreatic acinar membranes after subtotal enterectomy.
The American journal of physiology (1995-02-01)
I Alvaro-Alonso, B Colás, J P Esteve, C Susini, E Arilla
ABSTRACT
In the present study we found that exocrine pancreatic hyperplasia observed after proximal small bowel resection is accompanied by an increase in pancreatic somatostatin (SS) content at 1 mo and an increase in the number of SS receptors at 2 wk and 1 mo after intestinal surgery. At 6 mo after small bowel resection SS content and SS receptors had returned to control values. However, the original increase in SS receptor number was accompanied by a decrease in the ability of SS to inhibit forskolin-stimulated adenylyl cyclase (AC) activity. In addition, the ability of 5'-guanylylimidodiphosphate (a nonhydrolyzable GTP analogue) to inhibit SS receptor binding was decreased in pancreatic acinar membranes from enterectomized rats at 2 wk and 1 mo after jejunoileal resection. These data suggest that there is an abnormality in the integrity of SS receptor binding site-G protein interactions and would explain the decreased inactivation of AC by SS at 2 wk and 1 mo after proximal small bowel resection.