- Loss of hepatic monooxygenase activities, glutathione, and 'green pigment' formation after the administration of vinyl-cyclooctane to mice.
Loss of hepatic monooxygenase activities, glutathione, and 'green pigment' formation after the administration of vinyl-cyclooctane to mice.
Toxicology letters (1983-05-01)
P G Gervasi, L Citti, G Fassina, E Testai, G Turchi
PMID6857717
ABSTRACT
Vinylcyclooctane, when administered to mice at 500 mg/kg, produced reduction of microsomal cytochrome P-450, heme, aminopyrine-N-demethylase and ethoxycoumarin-O-deethylase activities with respect to control values; furthermore the hepatic reduced glutathione level was depleted suggesting that glutathione is involved in the vinylcyclooctane metabolism. The reduction of cytochrome P-450 and monooxygenase activities was accompanied by the formation of abnormal 'green pigments'.