Merck
  • The effect of methylbromophenvinfos (Polfos) on some enzymes in vivo and in vitro. I. In vivo studies.
The effect of methylbromophenvinfos (Polfos) on some enzymes in vivo and in vitro. I. In vivo studies.
Polish journal of pharmacology and pharmacy (1987-07-01)
D Pawłowska, D Koćmierska-Grodzka, J Moniuszko-Jakoniuk
ABSTRACT
The experiment was carried out on Wistar rats receiving orally either oil or oily solution of methylbromophenvinfos (Polfos) either in a single dose of 0.5 LD50, or doses of 0.1 LD50 once daily for a period of 2, 4 or 6 weeks. The activities of cholinesterase (ChE), beta-glucuronidase (beta-glu), lipase and amylase were assayed in the blood serum, the activity of acetylcholinesterase (AChE)-in brain homogenates, and the activities of lipase and amylase-in homogenates of the pancreas. Cholinesterases were inhibited in the course of both acute and chronic poisoning with Polfos. During the acute poisoning a sharp increase in the activity of beta-glu in the blood serum, 1 and 2 h after the pesticide administration, was observed. Polfos inhibited lipase and amylase both after acute and chronic treatment.