Merck
  • How can genipin assist gelatin/carbohydrate chitosan scaffolds to act as replacements of load-bearing soft tissues?
How can genipin assist gelatin/carbohydrate chitosan scaffolds to act as replacements of load-bearing soft tissues?
Carbohydrate polymers (2013-03-19)
Melika Sarem, Fathollah Moztarzadeh, Masoud Mozafari
ABSTRACT
In the engineering of soft tissues, designing the scaffolds with adequate strength and controllable biodegradation properties are essential. To fulfill such design criteria gelatin/carbohydrate chitosan (G/CC) scaffolds have gained much attention in engineering replacement of soft tissues. In this research, the influence of different cross-linking methods and parameters with genipin on the physico-chemical properties of G/CC scaffolds was investigated in detail. The scaffolds with different concentrations of G and CC were prepared and cross-linked trough two different methods, and the impact of cross-linker concentration beside of cross-linking time and temperature were fully analyzed. The obtained results suggested that the 1% genipin-cross-linked G60/CC40 scaffolds, prepared at room temperature for 24h through scaffold cross-linking method, could be a promising replacement in missing segments of load-bearing soft tissues. This article describes a systematic study of genipin cross-linking effects on G/CC scaffolds.
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology