Merck
Shotgun proteomic analysis of two Bartonella quintana strains.
Proteomics (2013-03-02)
Alessandra Fabietti, Marco Gaspari, Shibu Krishnan, Angela Quirino, Maria Carla Liberto, Giovanni Cuda, Alfredo Focà
ABSTRACT
This study reports the first extensive shotgun analysis of the Bartonella quintana proteome. Proteins extracted from two B. quintana strains, Oklahoma and JK31, were analyzed in triplicate analyses by a bottom-up approach consisting of tryptic digestion in SDS-containing buffer, strong cation-exchange StageTip fractionation, and nano-LC-MS/MS analysis. By setting spectral false discovery rate below 0.5%, 548 unique proteins were identified overall, of which 409 protein identifications were shared between the two strains. The data set, which achieves the highest proteome coverage for B. quintana to date, could be exploited for the quantitative analysis of a selected subset of target proteins.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O