Merck
  • Lovastatin corrects excess protein synthesis and prevents epileptogenesis in a mouse model of fragile X syndrome.
Lovastatin corrects excess protein synthesis and prevents epileptogenesis in a mouse model of fragile X syndrome.
Neuron (2013-01-29)
Emily K Osterweil, Shih-Chieh Chuang, Alexander A Chubykin, Michael Sidorov, Riccardo Bianchi, Robert K S Wong, Mark F Bear
ABSTRACT
Many neuropsychiatric symptoms of fragile X syndrome (FXS) are believed to be a consequence of altered regulation of protein synthesis at synapses. We discovered that lovastatin, a drug that is widely prescribed for the treatment of high cholesterol, can correct excess hippocampal protein synthesis in the mouse model of FXS and can prevent one of the robust functional consequences of increased protein synthesis in FXS, epileptogenesis. These data suggest that lovastatin is potentially disease modifying and could be a viable prophylactic treatment for epileptogenesis in FXS.
Product Description
Lovastatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Lovastatin for peak identification, European Pharmacopoeia (EP) Reference Standard