Merck
Pyrophosphate selective fluorescent probe and molecular flip-flop.
Dalton transactions (Cambridge, England : 2003) (2013-01-26)
Vijay Luxami, Kamaldeep Paul, In Howa Jeong
ABSTRACT
Highly selective detection of pyrophosphate in the presence of inorganic phosphates, halides, acetate, ATP, other nucleotides in neutral aqueous solutions is achieved by a fluorescent Zn-ensemble based system. The implementation of the first molecule based JK-latch sequential logic function is also described.
Sigma-Aldrich
Pyrophosphoric acid, SAJ first grade, ≥90.0%
Sigma-Aldrich
Pyrophosphoric acid, technical grade