Merck
  • Antioxidant capacity and inhibitory effect of grape seed and rosemary extract in marinades on the formation of heterocyclic amines in fried beef patties.
Antioxidant capacity and inhibitory effect of grape seed and rosemary extract in marinades on the formation of heterocyclic amines in fried beef patties.
Food chemistry (2012-10-31)
Monika Gibis, Jochen Weiss
ABSTRACT
The effect of oil-based marinades containing grape seed extract (Vitis vinifera L.; 0.2, 0.4, 0.6 and 0.8 g/100g) formulated in a water/oil emulsion or rosemary extract (Rosmarinus officinalis; 0.12, 0.2, 0.6, 1.0 and 1.5 g/100g) in oil on the formation of heterocyclic amines (HAs) in fried beef patties was examined. After application of marinades and frying, four HAs MeIQx (2-amino-3,8-dimethylimidazo[4,5-f]quinoxaline), PhIP (2-amino-1-methyl-6-phenylimidazo[4,5b]pyridine), Norharman, and Harman were found at low levels in all fried patties, MeIQx (0.3-1.0 ng/g), and PhIP (0.02-0.3 ng/g). The content of MeIQx and PhIP were significantly reduced by approx. 57% and 90% (p<0.05), respectively, after use of marinades containing the highest extract concentration. The antioxidant capacity of grape seed was about two-times greater than that of rosemary extract. A correlation between inhibition of HAs and Trolox-equivalents (MeIQx, R(2)=0.85, p<0.001; PhIP, R(2)=0.83, p<0.001) was found. Sensory tests showed a high acceptance of flavour and colour for controls and samples.