- Spatial structure of heptapeptide Aβ(16-22) (beta-amyloid Aβ(1-40) active fragment) in solution and in complex with a biological membrane model.
Spatial structure of heptapeptide Aβ(16-22) (beta-amyloid Aβ(1-40) active fragment) in solution and in complex with a biological membrane model.
Magnetic resonance in chemistry : MRC (2012-10-05)
Konstantin S Usachev, Sergej V Efimov, Ajdar R Yulmetov, Andrey V Filippov, Oleg N Antzutkin, Sergii Afonin, Vladimir V Klochkov
PMID23034896
ABSTRACT
The spatial structure of an active fragment of beta-amyloid Aβ(1-40) heptapeptide Aβ(16-22) (Lys-Leu-Val-Phe-Phe-Ala-Glu) in aqueous buffer solution and in complex with sodium dodecyl sulfate micelles as a model membrane system was investigated by (1)H NMR spectroscopy and two-dimensional NMR (TOCSY, HSQC-HECADE (Heteronuclear Couplings from ASSCI-domain experiments with E.COSY-type crosspeaks), NOESY) spectroscopy. Complex formation was confirmed by the chemical shift changes of the heptapeptide's (1)H NMR spectra, as well as by the signs and values of the NOE effects in different environments. We compared the spatial structure of the heptapeptide in borate buffer solution and in complex with a model of the cell surface membrane.