Merck
  • Phosphodiesterase type 5 inhibition reverts prostate fibroblast-to-myofibroblast trans-differentiation.
Phosphodiesterase type 5 inhibition reverts prostate fibroblast-to-myofibroblast trans-differentiation.
Endocrinology (2012-09-06)
Christoph Zenzmaier, Johann Kern, Natalie Sampson, Martin Heitz, Eugen Plas, Gerold Untergasser, Peter Berger
ABSTRACT
Phosphodiesterase type 5 (PDE5) inhibitors have been demonstrated to improve lower urinary tract symptoms secondary to benign prostatic hyperplasia (BPH). Because BPH is primarily driven by fibroblast-to-myofibroblast trans-differentiation, this study aimed to evaluate the potential of the PDE5 inhibitor vardenafil to inhibit and reverse trans-differentiation of primary human prostatic stromal cells (PrSC). Vardenafil, sodium nitroprusside, lentiviral-delivered short hairpin RNA-mediated PDE5 knockdown, sodium orthovanadate, and inhibitors of MAPK kinase, protein kinase G, Ras homolog family member (Rho) A, RhoA/Rho kinase, phosphatidylinositol 3 kinase and protein kinase B (AKT) were applied to PrSC treated with basic fibroblast growth factor (fibroblasts) or TGFβ1 (myofibroblasts) in vitro, in chicken chorioallantoic membrane xenografts in vivo, and to prostatic organoids ex vivo. Fibroblast-to-myofibroblast trans-differentiation was monitored by smooth muscle cell actin and IGF binding protein 3 mRNA and protein levels. Vardenafil significantly attenuated TGFβ1-induced PrSC trans-differentiation in vitro and in chorioallantoic membrane xenografts. Enhancement of nitric oxide/cyclic guanosine monophosphate signaling by vardenafil, sodium nitroprusside, or PDE5 knockdown reduced smooth muscle cell actin and IGF binding protein 3 mRNA and protein levels and restored fibroblast-like morphology in trans-differentiated myofibroblast. This reversal of trans-differentiation was not affected by MAPK kinase, protein kinase G, RhoA, or RhoA/Rho kinase inhibition, but vardenafil attenuated phospho-AKT levels in myofibroblasts. Consistently, phosphatidylinositol 3 kinase or AKT inhibition induced reversal of trans-differentiation, whereas the tyrosine phosphatase inhibitor sodium orthovanadate abrogated the effect of vardenafil. Treatment of prostatic organoids with vardenafil ex vivo reduced expression of myofibroblast markers, indicating reverse remodeling of stroma towards a desired higher fibroblast/myofibroblast ratio. Thus, enhancement of the nitric oxide/cyclic guanosine monophosphate signaling pathway by vardenafil attenuates and reverts fibroblast-to-myofibroblast trans-differentiation, hypothesizing that BPH patients might benefit from long-term therapy with PDE5 inhibitors.
Product Description
Supelco
Vardenafil dihydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Zopiclone-D4 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®