MilliporeSigma
  • Tryptophan-130 is the most reactive tryptophan residue in rabbit skeletal myosin subfragment-1.
Tryptophan-130 is the most reactive tryptophan residue in rabbit skeletal myosin subfragment-1.
FEBS letters (1990-01-01)
Y M Peyser, A Muhlrad, M M Werber
ABSTRACT
Rabbit skeletal muscle myosin subfragment-1 (S-1) was reacted with dimethyl(2-hydroxy-5-nitrobenzyl)sulfonium bromide (DHNBS) resulting in modification of 0.8 tryptophan residues per S-1. In order to assign the most reactive tryptophan of the 5 S-1 tryptophans, antibodies were raised in rabbits against bovine serum albumin modified with DHNBS. The antibodies reacted with the 27 kDa tryptic fragment of DHNBS-treated S-1, indicating that the reactive tryptophan resides on this domain. The 27 kDa fragment was isolated from DHNBS-treated S-1 and was further cleaved at a single cysteine residue by 2-nitro-5-thiocyanobenzoic acid. This cleavage resulted in two peptides, each of them containing one tryptophan. The antibodies reacted with the smaller peptide consisting of residues 122-204. The only tryptophan residing on this peptide is Trp130, and this is therefore the most reactive tryptophan of S-1.