Merck
  • Anaerobic transformation of octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX) by ovine rumen microorganisms.
Anaerobic transformation of octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX) by ovine rumen microorganisms.
Research in microbiology (2012-08-21)
Sudeep Perumbakkam, A M Craig
ABSTRACT
Explosives such as octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX) provide a challenge in terms of bioremediation. In the present study, sheep rumen was studied for its potential to detoxify HMX using analytical chemistry and molecular microbial ecology tools. Results indicated significant loss (p < 0.05) of HMX at 8 h post-incubation and complete disappearance of the parent molecule after 16 h. Qualitative LC-MS/MS analysis provided evidence for the formation of 1-NO-HMX and MEDINA metabolites. A total of 1006 16S rRNA-V3 clones were sequenced and the Classifier tool of the RDPII database was used to sort the sequences at their phylum level. Most sequences were associated with either the phylum Bacteroidetes or Firmicutes. Significant differences at the phylum level (p < 0.001) were found between 0 h and 8 h HMX treatments. Using LibCompare analysis, 8 h HMX treatment showed enrichment of clones (p < 0.01) belonging to the genus Prevotella. From these results, it could be concluded that members of the genus Prevotella are enriched in the rumen and are capable of detoxifying HMX.
Product Description
Supelco
HMX solution, 1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®