Merck
  • Phytoceramides and acylated phytosterol glucosides from Pterospermum acerifolium Willd. seed coat and their osteogenic activity.
Phytoceramides and acylated phytosterol glucosides from Pterospermum acerifolium Willd. seed coat and their osteogenic activity.
Phytochemistry (2012-07-13)
Preety Dixit, Kailash Chand, Mohd Parvez Khan, Jawed Akhtar Siddiqui, Deepshikha Tewari, Florence Tsofack Ngueguim, Naibedya Chattopadhyay, Rakesh Maurya
ABSTRACT
Phytochemical investigation of seed coats of Pterospermum acerifolium afforded two phytoceramides (1, 2) and two acylated phytosterol glucosides (3, 4) together with five known compounds (5-9). Their structures were elucidated on the basis of extensive spectroscopic analysis using 1D, 2D NMR and Mass spectrometry. Compounds 1, 2, 3, and 4 were assessed for their osteogenic activity using primary cultures of osteoblasts harvested from neonatal rat calvaria. Among these compounds, 1 and 2 markedly stimulated osteoblast differentiation assessed by alkaline phosphatase production and osteoblast mineralization by alizarin red-S staining.
Sigma-Aldrich
Alizarin Red S, SAJ special grade
Sigma-Aldrich
Alizarin Red S, certified by the BSC