Merck
  • A model study for constructing the DEF-benzoxocin ring system of menogaril and nogalamycin via a reductive Heck cyclization.
A model study for constructing the DEF-benzoxocin ring system of menogaril and nogalamycin via a reductive Heck cyclization.
Organic letters (2012-04-07)
Ruogu Peng, Michael S VanNieuwenhze
ABSTRACT
A novel reductive Heck cyclization approach was developed in order to construct a model DEF-benzoxocin ring system that is present in nogalamycin, menogaril, and related anthracycline antitumor antibiotics.