Merck
  • Histopathological evaluation of anterior lamellar corneal tissue-on/-off storage conditions on DSAEK donor tissue after storage in organ culture.
Histopathological evaluation of anterior lamellar corneal tissue-on/-off storage conditions on DSAEK donor tissue after storage in organ culture.
Current eye research (2012-01-19)
Vishal Jhanji, Graeme A Pollock, Adrienne L Mackey, Jacqueline Beltz, Rasik B Vajpayee
ABSTRACT
To compare the quality of corneal endothelium of precut Descemet-stripping automated endothelial keratoplasty (DSAEK) tissue when transported with and without the anterior lamellar corneal tissue (ALCT) when organ-culture corneal storage methods are used. After microkeratome-assisted excision of anterior corneal lamella, five pairs of corneas (10 eyes) were stored either with the ALCT on the stroma (five eyes) or with ALCT off the stroma (five eyes) in organ-culture medium. Three pairs (six matched corneas) were left in the transport medium for 24 h prior to the microkeratome cut. Two pairs (four matched corneas) were left in the transport medium for 48 h prior to the microkeratome cut. All cuts were performed using a 300 (four eyes) or 350 (six eyes) microns head. A vital dye assay (alizarin red S and trypan blue) was used to identify devitalized and necrotic endothelial cells. In all matched cases, there was no difference between the endothelial cell appearance with or without the anterior corneal lamella. In all cases, there was no evidence for trypan blue stained cells beyond that normally seen on acceptable transplantable corneas and there was no evidence of loss of cells or any lifting of Descemet's membrane. There is no difference between the quality of the donor endothelial cell appearance with ALCT-on or -off when the donor cornea is stored using the organ-culture system of corneal storage. Organ-culture storage system is a safe and effective system in regard to preparation and transport of donor lenticules for DSAEK.
Sigma-Aldrich
Alizarin Red S, certified by the Biological Stain Commission
Sigma-Aldrich
Alizarin Red S, SAJ special grade