Merck
  • Cross-linked DNA generated by "bis-click" reactions with bis-functional azides: site independent ligation of oligonucleotides via nucleobase alkynyl chains.
Cross-linked DNA generated by "bis-click" reactions with bis-functional azides: site independent ligation of oligonucleotides via nucleobase alkynyl chains.
The Journal of organic chemistry (2010-11-13)
Suresh S Pujari, Hai Xiong, Frank Seela
ABSTRACT
Template-free cross-linking of single-stranded DNA bearing octadiynyl side chains at the 7-position of 8-aza-7-deazapurine moieties with bisfunctional azides is reported employing a Cu(I)-catalyzed azide-alkyne "bis-click" reaction. Bis-adducts were formed when the bis-azide:oligonucleotide ratio was 1:1; monofunctionalization occurred when the ratio was 15:1. Four-stranded DNA consisting of two cross-linked duplexes was obtained after hydridization. Cross-linked duplexes are as stable as individual duplexes when ligation was introduced at terminal positions; ligation at a central position led to a slight duplex destabilization.
Sigma-Aldrich
4-Azabenzimidazole, 99%