- Oxytocin receptors in the nucleus accumbens shell are involved in the consolidation of maternal memory in postpartum rats.
Oxytocin receptors in the nucleus accumbens shell are involved in the consolidation of maternal memory in postpartum rats.
Female rats with maternal experience display a shorter onset of maternal responsiveness compared to those with no prior experience. This phenomenon called 'maternal memory' is critically dependent on the nucleus accumbens (NA) shell. We hypothesized that activation of OT receptors in the NA shell facilitates maternal memory. In Experiment 1, postpartum female rats given 1 hour of maternal experience were infused following the experience with either a high or low dose of an OT antagonist into the NA shell and tested for maternal behavior after a 10-day pup isolation period. Females receiving a high dose of the antagonist showed a significantly longer latency to exhibit full maternal behavior after the pup isolation period compared to females that received vehicle or a high dose of antagonist in a control region. In Experiment 2, postpartum female rats were infused with either a high or low dose of OT into the NA shell after a 15-minute maternal experience and tested for maternal behavior after a 10-day pup isolation period. There were no significant differences between the females infused with OT and females treated with a vehicle infused into the NA shell or with OT infused into the control region. One possible reason for a lack of facilitation is a floor effect, since females in the control groups displayed a rapid maternal response after the pup isolation period. These findings suggest that OT receptors, likely in combination with other neurotransmitters, in the NA shell play a role in the consolidation of maternal memory.