MilliporeSigma
  • [The determination of the oxidation phenotype in inbred C57Bl/6 and BALB/c mice].
[The determination of the oxidation phenotype in inbred C57Bl/6 and BALB/c mice].
Biulleten' eksperimental'noi biologii i meditsiny (1990-11-01)
S B Seredenin, I V Rybina, T G Khlopushina, V P Zherdev
ABSTRACT
Antipyrine oxidation was studied in C57BL/6 and BALB/c inbred mice. It was found that C57BL/6 are weak oxidant but BALB/c are strong oxidants of antipyrine. Animals F1 hybrids inherited the high capacity of antipyrine oxidation.
Sigma-Aldrich
4-Hydroxyantipyrine, 99%