Merck
  • Epitaxially driven formation of intricate supported gold nanostructures on a lattice-matched oxide substrate.
Epitaxially driven formation of intricate supported gold nanostructures on a lattice-matched oxide substrate.
Nano letters (2009-10-22)
Gabriel A Devenyi, Jianfeng Li, Robert A Hughes, An-Chang Shi, Peter Mascher, John S Preston
ABSTRACT
A new class of gold nanostructures has been fabricated on the (100), (111), and (110) surfaces of lattice-matched MgAl(2)O(4) substrates. The nanostructures were fabricated through a synthesis route where a thin gold film dewets, liquefies, and then slowly self-assembles. The supported nanostructures are intricately shaped, crystalline, and epitaxially aligned. Simulations based on a continuum elastic theory indicate that the self-assembly is driven by strained epitaxy and minimization of the surface free energy.
Sigma-Aldrich
Magnesium aluminate, spinel, nanopowder, <50 nm particle size (BET)