Merck
  • Synthesis of acrylic polymer beads for solid-supported proline-derived organocatalysts.
Synthesis of acrylic polymer beads for solid-supported proline-derived organocatalysts.
Organic letters (2009-06-16)
Tor E Kristensen, Kristian Vestli, Kim A Fredriksen, Finn K Hansen, Tore Hansen
ABSTRACT
A completely non-chromatographic and highly large-scale adaptable synthesis of acrylic polymer beads containing proline and prolineamides has been developed. Novel monomeric proline (meth)acrylates are prepared from hydroxyproline in only one step. Free-radical copolymerization then gives solid-supported proline organocatalysts directly in as little as two steps overall, without using any prefabricated solid supports, by using either droplet or dispersion polymerization. These affordable acrylic beads have highly favorable and adjustable swelling characteristics and are excellent reusable catalysts for organocatalytic reactions.
Sigma-Aldrich
L-Prolinamide, 98%