- Syntheses and activities of N-substituted derivatives of siastatin B.
Syntheses and activities of N-substituted derivatives of siastatin B.
The Journal of antibiotics (1992-10-01)
T Kudo, Y Nishimura, S Kondo, T Takeuchi
PMID1473994
ABSTRACT
N-Substituted derivatives of siastatin B have been obtained by a chemical modification. Some derivatives showed potent inhibitory activities against Streptococcus sp. and Clostridium perfringens neuraminidases.