- Mixed drug intoxication involving zaleplon ("Sonata").
Mixed drug intoxication involving zaleplon ("Sonata").
Forensic science international (2003-07-10)
Karla A Moore, Tasha L Zemrus, Vera Ramcharitar, Barry Levine, David R Fowler
PMID12850405
ABSTRACT
Zaleplon ("Sonata") is a pyrazolopyrimidine derivative approved for use in the United States for the treatment of insomnia. To date, there has been little data in the toxicological literature where zaleplon has been implicated as causing a fatal intoxication, either alone or in combination with other drugs. This report documents a case where zaleplon was identified in a suicide by multiple drug ingestion. The following zaleplon concentrations were found: heart blood 2.2mg/l; bile 8.6mg/l and urine 1.4mg/l. Zaleplon was also detected but not quantitated in the kidney and liver.
Product Description
Supelco
Butalbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®